Product Details
- SNP ID
-
rs2268242
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:33408798 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAAATATTTTTAAATGTTATAGGA[C/T]ACTACTTGCCAGGAAGAGTTGAAGA
- Phenotype
-
MIM: 147569
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IFNGR2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs144251244] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IFNGR2
- Gene Name
- interferon gamma receptor 2 (interferon gamma transducer 1)
View Full Product Details