Product Details
- SNP ID
-
rs2281068
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:129653796 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGCAGGGAGACTAAAGTGAAGCATG[C/T]CCGTGCCTGGGACAGAAAGGAAGGC
- Phenotype
-
MIM: 300297
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
APLN
PubMed Links
Gene Details
- Gene
- APLN
- Gene Name
- apelin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017413.4 |
|
Intron |
|
|
NP_059109.3 |
View Full Product Details