Product Details

SNP ID
rs2268992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:87473929 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATCTTCTTGCGTGGGTTTCAAGTT[A/G]TTTTCCCTGTTGCGGGGTCCATGTT
Phenotype
MIM: 605634
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFAP206 PubMed Links

Gene Details

Gene
CFAP206
Gene Name
cilia and flagella associated protein 206
There are no transcripts associated with this gene.

Gene
SLC35A1
Gene Name
solute carrier family 35 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168398.1 Intron NP_001161870.1
NM_006416.4 Intron NP_006407.1

View Full Product Details