Product Details

SNP ID
rs2174087
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:48102484 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TTGTGAGGGTCTAATAACTGTTATA[C/T]ACATTATCTCATTTAATCTTCACAA
Phenotype
MIM: 616392
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SKINT1L PubMed Links

Gene Details

Gene
SKINT1L
Gene Name
Skint1 like (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details