Product Details

SNP ID
rs2281492
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:161161316 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAACCCGCGGTGCAGCCATAGTCC[C/T]GTGAGCTGCCCACTTCCTGCCCTGG
Phenotype
MIM: 614795
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AGPAT4 PubMed Links

Gene Details

Gene
AGPAT4
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020133.2 Intron NP_064518.1
XM_005267053.3 Intron XP_005267110.1
XM_006715514.3 Intron XP_006715577.1
XM_017011059.1 Intron XP_016866548.1
Gene
AGPAT4-IT1
Gene Name
AGPAT4 intronic transcript 1
There are no transcripts associated with this gene.

View Full Product Details