Product Details

SNP ID
rs2269337
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:130697297 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCCATGTGCCACAGGAAAATAGTCC[A/G]TGTTTTTCATTGTCGTGTAATCGAA
Phenotype
MIM: 602238
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EXOSC2 PubMed Links

Gene Details

Gene
EXOSC2
Gene Name
exosome component 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282708.1 Intron NP_001269637.1
NM_001282709.1 Intron NP_001269638.1
NM_014285.6 Intron NP_055100.2
XM_005272176.2 Intron XP_005272233.1
XM_006717023.2 Intron XP_006717086.1
XM_017014558.1 Intron XP_016870047.1

View Full Product Details