Product Details

SNP ID
rs2269360
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:6884712 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GCCCAGACTCACTACCCGCCCTCTC[A/G]GTCCCGCAGCCACGAGGAGGACGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DSTNP2 PubMed Links

Gene Details

Gene
DSTNP2
Gene Name
destrin, actin depolymerizing factor pseudogene 2
There are no transcripts associated with this gene.

Gene
LOC105369632
Gene Name
uncharacterized LOC105369632
There are no transcripts associated with this gene.

Gene
RPL13P5
Gene Name
ribosomal protein L13 pseudogene 5
There are no transcripts associated with this gene.

View Full Product Details