Product Details

SNP ID
rs2282058
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:51936024 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGGAGGAGCGGGAGGAGAAAAGC[A/G]GCAGGAGCTGAGGGAGCCAGTTGCC
Phenotype
MIM: 606882
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP7B PubMed Links

Gene Details

Gene
ATP7B
Gene Name
ATPase copper transporting beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000053.3 Intron NP_000044.2
NM_001005918.2 Intron NP_001005918.1
NM_001243182.1 Intron NP_001230111.1
XM_005266423.2 Intron XP_005266480.1
XM_005266424.4 Intron XP_005266481.1
XM_005266427.2 Intron XP_005266484.1
XM_005266428.1 Intron XP_005266485.1
XM_005266430.4 Intron XP_005266487.1
XM_005266431.3 Intron XP_005266488.1
XM_005266432.2 Intron XP_005266489.1
XM_006719837.3 Intron XP_006719900.1
XM_006719838.1 Intron XP_006719901.1
XM_006719839.1 Intron XP_006719902.1
XM_011535117.2 Intron XP_011533419.1
XM_011535118.1 Intron XP_011533420.1
XM_011535119.1 Intron XP_011533421.1
XM_011535122.2 Intron XP_011533424.1
XM_017020627.1 Intron XP_016876116.1
XM_017020628.1 Intron XP_016876117.1

View Full Product Details