Product Details

SNP ID
rs2270048
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:2326216 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAACTTTGCAGCTGTAATTAGAGCC[C/T]CTGAATCAGCTGACTGAGCCAATCA
Phenotype
MIM: 607287 MIM: 608239
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LSM7 PubMed Links

Gene Details

Gene
LSM7
Gene Name
LSM7 homolog, U6 small nuclear RNA and mRNA degradation associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016199.2 Intron NP_057283.1
XM_017026869.1 Intron XP_016882358.1
Gene
SPPL2B
Gene Name
signal peptide peptidase like 2B
There are no transcripts associated with this gene.

View Full Product Details