Product Details

SNP ID
rs2282480
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63463904 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCCACTGAGGCCCTCATTCTACA[A/G]ACAGGGGATCCAAGAGCCAGGGAGC
Phenotype
MIM: 611474
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HRASLS5 PubMed Links

Gene Details

Gene
HRASLS5
Gene Name
HRAS like suppressor family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146728.1 Intron NP_001140200.1
NM_001146729.1 Intron NP_001140201.1
NM_054108.3 Intron NP_473449.1
XM_011544751.2 Intron XP_011543053.1
XM_011544752.2 Intron XP_011543054.1
XM_011544753.1 Intron XP_011543055.1
XM_017017172.1 Intron XP_016872661.1

View Full Product Details