Product Details

SNP ID
rs2288411
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8522805 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCAGGGGGCCCCCTCTGGCAGAG[A/G]GGGGCACCCCATTGCGATCCATGCC
Phenotype
MIM: 601480
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYO1F PubMed Links

Gene Details

Gene
MYO1F
Gene Name
myosin IF
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012335.3 2945 Missense Mutation CCC,CTC P960L NP_036467.2
XM_011528024.1 2945 Missense Mutation CCC,CTC P981L XP_011526326.1
XM_011528025.1 2945 Missense Mutation CCC,CTC P977L XP_011526327.1
XM_011528026.1 2945 Missense Mutation CCC,CTC P952L XP_011526328.1
XM_011528027.1 2945 Missense Mutation CCC,CTC P936L XP_011526329.1
XM_011528028.2 2945 Intron XP_011526330.1
XM_017026821.1 2945 Missense Mutation CCC,CTC P956L XP_016882310.1
Gene
ZNF414
Gene Name
zinc finger protein 414
There are no transcripts associated with this gene.

View Full Product Details