Product Details

SNP ID
rs2275404
Assay Type
Functionally tested
NCBI dbSNP Submissions
47
Location
Chr.1:160028650 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
AATATAATACTGTGGAACTTTTTTT[A/G]TCTTTTTGTGAGTGTTTTCTGACTG
Phenotype
MIM: 602208 MIM: 610273
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNJ10 PubMed Links

Gene Details

Gene
KCNJ10
Gene Name
potassium voltage-gated channel subfamily J member 10
There are no transcripts associated with this gene.

Gene
PIGM
Gene Name
phosphatidylinositol glycan anchor biosynthesis class M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145167.2 3344 UTR 3 NP_660150.1

View Full Product Details