Product Details

SNP ID
rs2288917
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40235212 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGAGGCCTCAACCAAGGTCACCA[C/T]GAGTGGGCCAGGTCCCTGAGGGTCC
Phenotype
MIM: 164731
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AKT2 PubMed Links

Gene Details

Gene
AKT2
Gene Name
AKT serine/threonine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243027.2 Intron NP_001229956.1
NM_001243028.2 Intron NP_001229957.1
NM_001626.5 Intron NP_001617.1
XM_011526614.1 Intron XP_011524916.1
XM_011526615.1 Intron XP_011524917.1
XM_011526616.1 Intron XP_011524918.1
XM_011526618.1 Intron XP_011524920.1
XM_011526619.1 Intron XP_011524921.1
XM_011526620.1 Intron XP_011524922.1
XM_011526622.2 Intron XP_011524924.1
XM_017026470.1 Intron XP_016881959.1
XM_017026471.1 Intron XP_016881960.1
Gene
CNTD2
Gene Name
cyclin N-terminal domain containing 2
There are no transcripts associated with this gene.

View Full Product Details