Product Details

SNP ID
rs2968166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:25721078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTGTACAATTCCAATTGCTCTGA[C/T]CATATTTTTATTTTCAAGTGTATTA
Phenotype
MIM: 610661
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NGLY1 PubMed Links

Gene Details

Gene
NGLY1
Gene Name
N-glycanase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145293.1 Intron NP_001138765.1
NM_001145294.1 Intron NP_001138766.1
NM_001145295.1 Intron NP_001138767.1
NM_018297.3 Intron NP_060767.2
XM_005265316.1 Intron XP_005265373.1
XM_005265317.1 Intron XP_005265374.1
XM_011533944.1 Intron XP_011532246.1
XM_011533945.2 Intron XP_011532247.1
XM_017006839.1 Intron XP_016862328.1

View Full Product Details