Product Details

SNP ID
rs2276064
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118658994 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTGGAAAGCCACTTCCTGGGCCC[A/G]ACGTAAAGGTGCCTTGGCCAGGCCT
Phenotype
MIM: 612834 MIM: 275360
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PHLDB1 PubMed Links

Gene Details

Gene
PHLDB1
Gene Name
pleckstrin homology like domain family B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144758.2 1501 Intron NP_001138230.1
NM_001144759.2 1501 Intron NP_001138231.1
NM_015157.3 1501 Intron NP_055972.1
XM_005271468.3 1501 Intron XP_005271525.1
XM_005271469.3 1501 Intron XP_005271526.1
XM_005271470.3 1501 Intron XP_005271527.1
XM_005271471.3 1501 Intron XP_005271528.1
XM_005271472.3 1501 Intron XP_005271529.1
XM_005271473.3 1501 Intron XP_005271530.1
XM_005271474.4 1501 Intron XP_005271531.1
XM_005271475.3 1501 Intron XP_005271532.1
XM_005277680.3 1501 Intron XP_005277737.1
XM_006718796.3 1501 Intron XP_006718859.1
XM_006718797.3 1501 Intron XP_006718860.1
XM_006718798.3 1501 Intron XP_006718861.1
XM_006718799.3 1501 Intron XP_006718862.1
XM_006718800.3 1501 Intron XP_006718863.1
XM_006718801.3 1501 Intron XP_006718864.1
XM_006718802.3 1501 Intron XP_006718865.1
XM_006718803.3 1501 Intron XP_006718866.1
XM_011542703.2 1501 Intron XP_011541005.1
XM_011542704.2 1501 Intron XP_011541006.1
XM_011542705.2 1501 Intron XP_011541007.1
XM_011542706.2 1501 Intron XP_011541008.1
XM_011542707.2 1501 Intron XP_011541009.1
XM_011542708.2 1501 Intron XP_011541010.1
XM_011542709.2 1501 Intron XP_011541011.1
XM_011542710.2 1501 Intron XP_011541012.1
XM_011542711.2 1501 Intron XP_011541013.1
XM_011542713.2 1501 Intron XP_011541015.1
XM_011542714.2 1501 Intron XP_011541016.1
XM_011542715.2 1501 Intron XP_011541017.1
XM_017017407.1 1501 Intron XP_016872896.1
XM_017017408.1 1501 Intron XP_016872897.1
XM_017017409.1 1501 Intron XP_016872898.1
XM_017017410.1 1501 Intron XP_016872899.1
XM_017017411.1 1501 Intron XP_016872900.1
Gene
TREH
Gene Name
trehalase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301065.1 1501 Missense Mutation CGG,TGG R455W NP_001287994.1
NM_007180.2 1501 Missense Mutation CGG,TGG R486W NP_009111.2

View Full Product Details