Product Details

SNP ID
rs2276118
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:67521123 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTCCCGGCAGCCAATGTAGCCGTCC[T/C]GGTCTCGGTCAAACTCCTGGAAGGC
Phenotype
MIM: 607314
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
CABP2 PubMed Links

Gene Details

Gene
CABP2
Gene Name
calcium binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318496.1 387 Missense Mutation CAG,CGG Q100R NP_001305425.1
NM_016366.2 387 Missense Mutation CAG,CGG Q94R NP_057450.2

View Full Product Details