Product Details

SNP ID
rs2968917
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:40583566 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGTGTTAATGCTTTGGGTTTTGC[C/T]TCAGAGTCTCTGGGCAGTGAGAGAA
Phenotype
MIM: 300556
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP6AP2 PubMed Links

Gene Details

Gene
ATP6AP2
Gene Name
ATPase H+ transporting accessory protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005765.2 Intron NP_005756.2

View Full Product Details