Product Details
- SNP ID
-
rs2276427
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:62243472 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGAAACGAAGCCTCATTGCGGAAG[C/T]CCTGGTAACTTCTTTCTCCTTTATT
- Phenotype
-
MIM: 615061
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SCGB1D2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2232951] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SCGB1D2
- Gene Name
- secretoglobin family 1D member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006551.3 |
295 |
Missense Mutation |
GCC,GTC |
A80V |
NP_006542.1 |
View Full Product Details