Product Details

SNP ID
rs2276531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44026606 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGTGGATTGCAGCTTTCTTTTGT[C/T]AATAGGCTTAAAGAATTATTATTTC
Phenotype
MIM: 602103
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRAPPC10 PubMed Links

Gene Details

Gene
TRAPPC10
Gene Name
trafficking protein particle complex 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003274.4 Intron NP_003265.3
XM_005261168.4 Intron XP_005261225.1
XM_011529714.2 Intron XP_011528016.1
XM_011529715.2 Intron XP_011528017.1
XM_011529716.2 Intron XP_011528018.1
XM_011529717.2 Intron XP_011528019.1
XM_011529718.2 Intron XP_011528020.1
XM_011529719.2 Intron XP_011528021.1
XM_011529720.2 Intron XP_011528022.1
XM_011529721.2 Intron XP_011528023.1
XM_011529722.2 Intron XP_011528024.1
XM_011529723.2 Intron XP_011528025.1
XM_011529725.2 Intron XP_011528027.1
XM_011529726.2 Intron XP_011528028.1
XM_011529727.2 Intron XP_011528029.1
XM_011529728.2 Intron XP_011528030.1
XM_011529729.2 Intron XP_011528031.1
XM_011529730.2 Intron XP_011528032.1
XM_017028454.1 Intron XP_016883943.1
XM_017028455.1 Intron XP_016883944.1

View Full Product Details