Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001001344.2 | Intron | NP_001001344.1 | |||
NM_021949.3 | Intron | NP_068768.2 | |||
XM_005274689.3 | Intron | XP_005274746.1 | |||
XM_005274690.3 | Intron | XP_005274747.1 | |||
XM_005274691.3 | Intron | XP_005274748.1 | |||
XM_005274692.3 | Intron | XP_005274749.1 | |||
XM_011531174.2 | Intron | XP_011529476.1 | |||
XM_011531177.2 | Intron | XP_011529479.1 | |||
XM_017029553.1 | Intron | XP_016885042.1 |