Product Details

SNP ID
rs2289711
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:27524572 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCTGGGGCTTGGACCGAGCTTGTAC[G/T]TGAAGAATACTCCAGGATTGGACGT
Phenotype
MIM: 603141
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PPFIBP1 PubMed Links

Gene Details

Gene
PPFIBP1
Gene Name
PPFIA binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198915.1 Intron NP_001185844.1
NM_001198916.1 Intron NP_001185845.1
NM_003622.3 Intron NP_003613.3
NM_177444.2 Intron NP_803193.2
XM_005253505.1 Intron XP_005253562.1
XM_005253507.1 Intron XP_005253564.1
XM_005253508.1 Intron XP_005253565.1
XM_005253509.1 Intron XP_005253566.1
XM_006719159.3 Intron XP_006719222.1
XM_017020056.1 Intron XP_016875545.1
XM_017020057.1 Intron XP_016875546.1
XM_017020058.1 Intron XP_016875547.1
XM_017020059.1 Intron XP_016875548.1
XM_017020060.1 Intron XP_016875549.1
XM_017020061.1 Intron XP_016875550.1
XM_017020062.1 Intron XP_016875551.1
XM_017020063.1 Intron XP_016875552.1
XM_017020064.1 Intron XP_016875553.1
XM_017020065.1 Intron XP_016875554.1
XM_017020066.1 Intron XP_016875555.1
XM_017020067.1 Intron XP_016875556.1
XM_017020068.1 Intron XP_016875557.1
XM_017020069.1 Intron XP_016875558.1
XM_017020070.1 Intron XP_016875559.1
XM_017020071.1 Intron XP_016875560.1
XM_017020072.1 Intron XP_016875561.1
XM_017020073.1 Intron XP_016875562.1
XM_017020074.1 Intron XP_016875563.1
XM_017020075.1 Intron XP_016875564.1
XM_017020076.1 Intron XP_016875565.1
XM_017020077.1 Intron XP_016875566.1
XM_017020078.1 Intron XP_016875567.1

View Full Product Details