Product Details

SNP ID
rs2276871
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:40457782 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGGCTCTCGGGCGTTTGTTCCCTGC[A/G]GCATTCTTTTTCGCTGAATTTAACC
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ENTPD3-AS1 PubMed Links

Gene Details

Gene
ENTPD3-AS1
Gene Name
ENTPD3 antisense RNA 1
There are no transcripts associated with this gene.

Gene
RPL14
Gene Name
ribosomal protein L14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034996.2 Intron NP_001030168.1
NM_003973.4 Intron NP_003964.3

View Full Product Details