Product Details

SNP ID
rs2276869
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:40457429 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCTCGACCTCCACCTCCGCTGGG[A/T]AGCTGAGGCGCCAAACGGCTCCCAG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ENTPD3-AS1 PubMed Links

Gene Details

Gene
ENTPD3-AS1
Gene Name
ENTPD3 antisense RNA 1
There are no transcripts associated with this gene.

Gene
RPL14
Gene Name
ribosomal protein L14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034996.2 91 UTR 5 NP_001030168.1
NM_003973.4 91 Intron NP_003964.3

View Full Product Details