Product Details

SNP ID
rs2277706
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:81843795 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GGAAACAAGCCCCACCAGGGTGGGC[G/T]GCCTGGAGATGGATCCCTTTCCAAA
Phenotype
MIM: 176790
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
P4HB PubMed Links

Gene Details

Gene
P4HB
Gene Name
prolyl 4-hydroxylase subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000918.3 1941 UTR 3 NP_000909.2
Gene
PPP1R27
Gene Name
protein phosphatase 1 regulatory subunit 27
There are no transcripts associated with this gene.

View Full Product Details