Product Details
- SNP ID
-
rs2519168
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:2947870 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGCCGCTGAAACACACTCGACCAT[A/T]GCTGCGCTGGCGAAGAAGCCCCAAG
- Phenotype
-
MIM: 601651
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NAP1L4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs16928844] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NAP1L4
- Gene Name
- nucleosome assembly protein 1 like 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005969.3 |
|
Intron |
|
|
NP_005960.1 |
View Full Product Details