Product Details

SNP ID
rs1801182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:94398387 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGATTTATTCTTTTCTAGGAAAC[C/T]GTAAGAAAGGTAAGAGTACACTACT
Phenotype
MIM: 120160
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COL1A2 PubMed Links

Gene Details

Gene
COL1A2
Gene Name
collagen type I alpha 2 chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000089.3 558 Silent Mutation ACC,ACT T29T NP_000080.2

View Full Product Details