Product Details

SNP ID
rs2520158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:10700744 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCACCACTGCACTCCAGCTTGCGCA[A/G]TAGAGCCAGACCCTGTCTCAGGAAA
Phenotype
MIM: 603644
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADPRM PubMed Links

Gene Details

Gene
ADPRM
Gene Name
ADP-ribose/CDP-alcohol diphosphatase, manganese dependent
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020233.4 Intron NP_064618.3
XM_005256738.2 Intron XP_005256795.1
Gene
SCO1
Gene Name
SCO1 cytochrome c oxidase assembly protein
There are no transcripts associated with this gene.

View Full Product Details