Product Details

SNP ID
rs2245910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:564538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGAAGTCCCTTAGACCAATAACA[A/T]AAAAATCACTTTTTAACTCAGGTGC
Phenotype
MIM: 612220 MIM: 607297
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
B4GALNT3 PubMed Links

Gene Details

Gene
B4GALNT3
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 3
There are no transcripts associated with this gene.

Gene
NINJ2
Gene Name
ninjurin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001294345.1 972 UTR 3 NP_001281274.1
NM_001294346.1 972 UTR 3 NP_001281275.1
NM_016533.5 972 UTR 3 NP_057617.2

View Full Product Details