Product Details
- SNP ID
-
rs2536192
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:121327159 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGCCGTTAGCCAAAATATTTCTGG[A/T]TCAGTATCTACATCCTATCATCAAC
- Phenotype
-
MIM: 606267
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
WNT16
PubMed Links
Gene Details
- Gene
- WNT16
- Gene Name
- Wnt family member 16
View Full Product Details