Product Details
- SNP ID
-
rs2497190
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:19517703 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAAGTCAGTTCTGTAATAAGAACT[C/G]TGCGTCCATGGCAACCCCAAACAGA
- Phenotype
-
MIM: 606791
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TPTE2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116286761] are located under a probe and SNP(s) [rs145197933] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TPTE2
- Gene Name
- transmembrane phosphoinositide 3-phosphatase and tensin homolog 2
View Full Product Details