Product Details

SNP ID
rs183561682
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:15070940 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTACATACAGGGTTTTATATATA[G/A]AAAAGGAGACACTTTATTCTAAAAT
Phenotype
MIM: 614244 MIM: 605121
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
PDXDC1 PubMed Links

Gene Details

Gene
PDXDC1
Gene Name
pyridoxal dependent decarboxylase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001285444.1 Intron NP_001272373.1
NM_001285445.1 Intron NP_001272374.1
NM_001285447.1 Intron NP_001272376.1
NM_001285448.1 Intron NP_001272377.1
NM_001285449.1 Intron NP_001272378.1
NM_001285450.1 Intron NP_001272379.1
NM_001324019.1 Intron NP_001310948.1
NM_001324020.1 Intron NP_001310949.1
NM_001324021.1 Intron NP_001310950.1
NM_015027.3 Intron NP_055842.2
XM_005255173.1 Intron XP_005255230.1
XM_005255176.2 Intron XP_005255233.1
XM_006720865.2 Intron XP_006720928.2
XM_017023059.1 Intron XP_016878548.1
XM_017023060.1 Intron XP_016878549.1
XM_017023061.1 Intron XP_016878550.1
XM_017023062.1 Intron XP_016878551.1
XM_017023063.1 Intron XP_016878552.1
XM_017023064.1 Intron XP_016878553.1
XM_017023065.1 Intron XP_016878554.1
Gene
RRN3
Gene Name
RRN3 homolog, RNA polymerase I transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301064.1 Intron NP_001287993.1
NM_018427.4 Intron NP_060897.3
XM_005255376.1 Intron XP_005255433.1
XM_005255377.4 Intron XP_005255434.1
XM_011522540.2 Intron XP_011520842.1
XM_011522541.1 Intron XP_011520843.1

View Full Product Details