Product Details

SNP ID
rs2507815
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65028300 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGAACTGGTAACAAGGAAGGTCC[A/C]CAGGGAGAGGAACTGTGTCACTGGG
Phenotype
MIM: 601175 MIM: 605964
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARL2 PubMed Links

Gene Details

Gene
ARL2
Gene Name
ADP ribosylation factor like GTPase 2
There are no transcripts associated with this gene.

Gene
ARL2-SNX15
Gene Name
ARL2-SNX15 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
MIR6879
Gene Name
microRNA 6879
There are no transcripts associated with this gene.

Gene
SNX15
Gene Name
sorting nexin 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013306.4 Intron NP_037438.2
NM_147777.3 Intron NP_680086.2

View Full Product Details