Product Details
- SNP ID
-
rs2551352
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:23691910 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGGGCCACATATGTCGCTTGGGGG[G/T]AAGAGTGCAGATGGGCGGAGAACTC
- Phenotype
-
MIM: 615347
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ATAD2B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78748802] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATAD2B
- Gene Name
- ATPase family, AAA domain containing 2B
- Gene
- KLHL29
- Gene Name
- kelch like family member 29
View Full Product Details