Product Details

SNP ID
rs2578662
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:45594986 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTTCCGAGTGGACAAGGGTGCAG[A/G]CAACAACCCCGAGTTTGAGGAAACT
Phenotype
MIM: 604543
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LIMD1 PubMed Links

Gene Details

Gene
LIMD1
Gene Name
LIM domains containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014240.2 156 Missense Mutation GAC,GGC D36G NP_055055.1
XM_011534207.2 156 Missense Mutation GAC,GGC D36G XP_011532509.1

View Full Product Details