Product Details

SNP ID
rs2243934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:127648346 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATATTATTAGGTAGTTGCCCTCC[C/T]CTTTGCTTGAGGACATTGATGGCCA
Phenotype
MIM: 603071 MIM: 607908
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR17 PubMed Links

Gene Details

Gene
GPR17
Gene Name
G protein-coupled receptor 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161415.1 Intron NP_001154887.1
NM_001161416.1 Intron NP_001154888.1
NM_001161417.1 Intron NP_001154889.1
NM_005291.2 Intron NP_005282.1
XM_017003833.1 Intron XP_016859322.1
Gene
LIMS2
Gene Name
LIM zinc finger domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136037.2 Intron NP_001129509.2
NM_001161403.1 Intron NP_001154875.1
NM_001161404.1 Intron NP_001154876.1
NM_001256542.1 Intron NP_001243471.1
NM_017980.4 Intron NP_060450.2
XM_005263710.2 Intron XP_005263767.1
XM_006712627.3 Intron XP_006712690.1
XM_006712628.3 Intron XP_006712691.1
XM_011511453.1 Intron XP_011509755.1
XM_017004469.1 Intron XP_016859958.1

View Full Product Details