Product Details

SNP ID
rs2588453
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:38349991 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCTCCAAATAATCACAATCCAAGG[A/C]CCTCGGGGTTAGGATTTCAGCATAT
Phenotype
MIM: 604049
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
XYLB PubMed Links

Gene Details

Gene
XYLB
Gene Name
xylulokinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005108.3 Intron NP_005099.2
XM_011534325.2 Intron XP_011532627.1
XM_011534327.1 Intron XP_011532629.1
XM_011534328.2 Intron XP_011532630.1
XM_011534329.1 Intron XP_011532631.1
XM_011534330.2 Intron XP_011532632.1
XM_017007595.1 Intron XP_016863084.1
XM_017007596.1 Intron XP_016863085.1
XM_017007597.1 Intron XP_016863086.1
XM_017007598.1 Intron XP_016863087.1
XM_017007599.1 Intron XP_016863088.1
XM_017007600.1 Intron XP_016863089.1

View Full Product Details