Product Details

SNP ID
rs2665646
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39639525 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGCAGAGGCTGGAGTGGGCTCA[G/T]AGATCAGAAAGCTGGGTCCTCTGAA
Phenotype
MIM: 604399 MIM: 607048
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PPP1R1B PubMed Links

Gene Details

Gene
PPP1R1B
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 1B
There are no transcripts associated with this gene.

Gene
STARD3
Gene Name
StAR related lipid transfer domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165937.1 Intron NP_001159409.1
NM_001165938.1 Intron NP_001159410.1
NM_006804.3 Intron NP_006795.3
XM_006721646.2 Intron XP_006721709.1
XM_011524210.1 Intron XP_011522512.1
XM_011524211.2 Intron XP_011522513.1
XM_011524213.1 Intron XP_011522515.1
XM_017024041.1 Intron XP_016879530.1

View Full Product Details