Product Details

SNP ID
rs2677931
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:6634518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCGGTTCCCCTTTTGGTATCTT[C/T]GGTGATGGTGGAGCTGTTTTCTCCC
Phenotype
MIM: 610916 MIM: 184753
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NSUN2 PubMed Links

Gene Details

Gene
NSUN2
Gene Name
NOP2/Sun RNA methyltransferase family member 2
There are no transcripts associated with this gene.

Gene
SRD5A1
Gene Name
steroid 5 alpha-reductase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001047.3 Intron NP_001038.1
NM_001324322.1 Intron NP_001311251.1
NM_001324323.1 Intron NP_001311252.1

View Full Product Details