Product Details

SNP ID
hCV15916591
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:70566993 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTAGCTGGGTCTCGGCCACGGCGT[G/T]GAGAGCGGCAAGTAAGCACCTGGCC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM135A PubMed Links

Gene Details

Gene
FAM135A
Gene Name
family with sequence similarity 135 member A
There are no transcripts associated with this gene.

Gene
SDHAF4
Gene Name
succinate dehydrogenase complex assembly factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145267.2 73 Missense Mutation TGG,TTG W18L NP_660310.2

View Full Product Details