Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_017573.4 | 2037 | UTR 3 | NP_060043.2 | ||
XM_005259586.1 | 2037 | UTR 3 | XP_005259643.1 | ||
XM_011528085.2 | 2037 | UTR 3 | XP_011526387.1 | ||
XM_011528086.2 | 2037 | UTR 3 | XP_011526388.1 | ||
XM_011528087.2 | 2037 | UTR 3 | XP_011526389.1 | ||
XM_011528088.2 | 2037 | UTR 3 | XP_011526390.1 | ||
XM_011528089.2 | 2037 | UTR 3 | XP_011526391.1 | ||
XM_011528090.2 | 2037 | UTR 3 | XP_011526392.1 | ||
XM_011528091.2 | 2037 | UTR 3 | XP_011526393.1 | ||
XM_011528092.2 | 2037 | Intron | XP_011526394.1 | ||
XM_011528093.2 | 2037 | UTR 3 | XP_011526395.1 | ||
XM_011528094.1 | 2037 | UTR 3 | XP_011526396.1 | ||
XM_011528095.2 | 2037 | Intron | XP_011526397.1 | ||
XM_011528096.1 | 2037 | UTR 3 | XP_011526398.1 | ||
XM_017026897.1 | 2037 | Intron | XP_016882386.1 |