Product Details

SNP ID
hCV15918930
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218578185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTATGATTGATTCCTTGAGGACCA[G/T]CCCTGAAGGGAGTTACAAAACAGCT
Phenotype
MIM: 612054
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CNOT9 PubMed Links

Gene Details

Gene
CNOT9
Gene Name
CCR4-NOT transcription complex subunit 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271634.1 Intron NP_001258563.1
NM_001271635.1 Intron NP_001258564.1
NM_005444.2 Intron NP_005435.1
Gene
USP37
Gene Name
ubiquitin specific peptidase 37
There are no transcripts associated with this gene.

View Full Product Details