Product Details
- SNP ID
-
rs2744389
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:7542890 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGACCCCAGTGTGCCTTGGATCGC[A/C]GAGCGCCGGCCGCATGTCTGCTTTG
- Phenotype
-
MIM: 125647
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DSP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76092821] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DSP
- Gene Name
- desmoplakin
- Gene
- LOC101928076
- Gene Name
- uncharacterized LOC101928076
There are no transcripts associated with this gene.
View Full Product Details