Product Details

SNP ID
rs2702022
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125595186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTTTTTTTTTTTTTTTCCTTTT[A/T]AAAGAGAGGAGGTCTGGCTCTATTG
Phenotype
MIM: 601134
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
STT3A PubMed Links

Gene Details

Gene
STT3A
Gene Name
STT3A, catalytic subunit of the oligosaccharyltransferase complex
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278503.1 Intron NP_001265432.1
NM_001278504.1 Intron NP_001265433.1
NM_152713.4 Intron NP_689926.1
XM_011542807.2 Intron XP_011541109.1
Gene
STT3A-AS1
Gene Name
STT3A antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details