Product Details

SNP ID
rs2075836
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:6951200 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACGAACTAGTTAGTATGTCTCTCTG[G/T]ATTTTAAACATAGTGAATTTCACAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DEFA8P PubMed Links

Gene Details

Gene
DEFA8P
Gene Name
defensin alpha 8, pseudogene
There are no transcripts associated with this gene.

Gene
DEFA9P
Gene Name
defensin alpha 9, pseudogene
There are no transcripts associated with this gene.

View Full Product Details