Product Details

SNP ID
rs2746122
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:20053733 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACCATATTATTTAGATATTTTAT[C/T]TCACTTTTCAAAATTTAAATAGAGA
Phenotype
MIM: 610952
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CFAP61 PubMed Links

Gene Details

Gene
CFAP61
Gene Name
cilia and flagella associated protein 61
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167816.1 Intron NP_001161288.1
NM_015585.3 Intron NP_056400.3
XM_005260688.3 Intron XP_005260745.1
XM_005260689.2 Intron XP_005260746.1
XM_005260690.3 Intron XP_005260747.1
XM_005260691.3 Intron XP_005260748.1
XM_005260692.3 Intron XP_005260749.1
XM_005260693.3 Intron XP_005260750.1
XM_005260695.4 Intron XP_005260752.1
XM_005260696.4 Intron XP_005260753.1
XM_005260697.1 Intron XP_005260754.1
XM_011529211.1 Intron XP_011527513.1
XM_011529212.2 Intron XP_011527514.1
XM_011529213.1 Intron XP_011527515.1
XM_017027786.1 Intron XP_016883275.1
XM_017027787.1 Intron XP_016883276.1
XM_017027788.1 Intron XP_016883277.1
XM_017027789.1 Intron XP_016883278.1
XM_017027790.1 Intron XP_016883279.1
XM_017027791.1 Intron XP_016883280.1
XM_017027792.1 Intron XP_016883281.1
XM_017027793.1 Intron XP_016883282.1
XM_017027794.1 Intron XP_016883283.1
XM_017027795.1 Intron XP_016883284.1
Gene
CRNKL1
Gene Name
crooked neck pre-mRNA splicing factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278625.1 Intron NP_001265554.1
NM_001278626.1 Intron NP_001265555.1
NM_001278627.1 Intron NP_001265556.1
NM_001278628.1 Intron NP_001265557.1
NM_016652.5 Intron NP_057736.4

View Full Product Details