Product Details

SNP ID
rs2763940
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:99614987 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCAGAATGTTTGAGTGGCACGAGT[A/G]CTTCATTCGTCTAGAAATATTTCAT
Phenotype
MIM: 609686
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLYBL PubMed Links

Gene Details

Gene
CLYBL
Gene Name
citrate lyase beta like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206808.3 Intron NP_996531.1
XM_005254030.2 Intron XP_005254087.1
XM_005254031.2 Intron XP_005254088.1
XM_006719915.3 Intron XP_006719978.1
XM_011521051.2 Intron XP_011519353.1
XM_011521052.2 Intron XP_011519354.1
XM_017020402.1 Intron XP_016875891.1
XM_017020403.1 Intron XP_016875892.1
XM_017020404.1 Intron XP_016875893.1

View Full Product Details