Product Details

SNP ID
rs2760235
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:117916524 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCCTGAAAATCTGTACCTATAA[A/G]GTGATGATGTGGAAGCAGTAGAGTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC35F1 PubMed Links

Gene Details

Gene
SLC35F1
Gene Name
solute carrier family 35 member F1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029858.3 Intron NP_001025029.2
XM_005266865.4 Intron XP_005266922.1

View Full Product Details