Product Details

SNP ID
rs2760257
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:130865910 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GATAAAGATCATCTTTATGTTGATT[G/T]AAAAATTTTTTAACTTTGTTTAATC
Phenotype
MIM: 604384
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ATP2C1 PubMed Links

Gene Details

Gene
ATP2C1
Gene Name
ATPase secretory pathway Ca2+ transporting 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001485.2 Intron NP_001001485.1
NM_001001486.1 Intron NP_001001486.1
NM_001001487.1 Intron NP_001001487.1
NM_001199179.1 Intron NP_001186108.1
NM_001199180.1 Intron NP_001186109.1
NM_001199181.1 Intron NP_001186110.1
NM_001199182.1 Intron NP_001186111.1
NM_001199183.1 Intron NP_001186112.1
NM_001199184.1 Intron NP_001186113.1
NM_001199185.1 Intron NP_001186114.1
NM_014382.3 Intron NP_055197.2
XM_005247354.2 Intron XP_005247411.1
XM_005247355.2 Intron XP_005247412.1
XM_005247356.2 Intron XP_005247413.1
XM_005247358.2 Intron XP_005247415.1
XM_011512686.2 Intron XP_011510988.1
XM_017006164.1 Intron XP_016861653.1

View Full Product Details