Product Details

SNP ID
rs2760352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:46776504 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTAGTTCCAGTTCTGGTTCCACCA[C/G]TTACTAGCTATGTGGCCTTAGATAA
Phenotype
MIM: 133280
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ESD PubMed Links

Gene Details

Gene
ESD
Gene Name
esterase D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001984.1 Intron NP_001975.1
XM_005266278.2 Intron XP_005266335.1
XM_011534954.1 Intron XP_011533256.1
XM_017020414.1 Intron XP_016875903.1

View Full Product Details