Product Details

SNP ID
rs2760533
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:192578911 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTCATGTCTGCTTCTTTTGCCT[C/T]TCCTAACTATACTATGAGGAAATTT
Phenotype
MIM: 600323
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RGS1 PubMed Links

Gene Details

Gene
RGS1
Gene Name
regulator of G-protein signaling 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002922.3 Intron NP_002913.3

View Full Product Details